Stampa…

2-Methoxy-4-nitrobenzaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

M2656-1G M2656-5G
TCIAM2656-1GEA 65 EUR
TCIAM2656-1G TCIAM2656-5G
2-Methoxy-4-nitrobenzaldehyde ≥98.0% (by GC)
2-Methoxy-4-nitrobenzaldehyde
Formula: C₈H₇NO₄
Peso molecolare: 181,15 g/mol
MDL Number: MFCD02683052
CAS Number: 136507-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e06d7c229d519bf0e2f57fbd1ddc553b7c8708c1&vid=e06d7c229d519bf0e2f57fbd1ddc553b7c8708c1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR