Stampa…

Methyl-3-amino-6-bromopyrazine-2-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 3-amino-6-bromopyrazine-2-carboxylate

M2655-1G M2655-5G
TCIAM2655-1GEA 40 EUR
TCIAM2655-1G TCIAM2655-5G
Methyl-3-amino-6-bromopyrazine-2-carboxylate ≥98.0% (by GC)
Methyl-3-amino-6-bromopyrazine-2-carboxylate
Formula: C₆H₆BrN₃O₂
Peso molecolare: 232,03 g/mol
MDL Number: MFCD00834964
CAS Number: 6966-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae5fb2829e5d292730c9e995d4c2904c42c1fcb0&vid=ae5fb2829e5d292730c9e995d4c2904c42c1fcb0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 172.0 to 176.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR