Stampa…

N-Methyl paroxetine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

M2645-200MG M2645-1G
TCIAM2645-200MGEA 54 EUR
TCIAM2645-200MG TCIAM2645-1G
N-Methyl paroxetine ≥98.0% (by GC, titration analysis)
N-Methyl paroxetine
Formula: C₂₀H₂₂FNO₃
Peso molecolare: 343,40 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD03788781
CAS Number: 110429-36-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c9850eb1bd712ef2d549ea0b975465dfd34e2c8&vid=9c9850eb1bd712ef2d549ea0b975465dfd34e2c8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Specific rotation [a]20/D -69.0 to -89.0 deg (C=0.01, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR