Stampa…

Methyl-6-chloropicolinate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 6-chloropyridine-2-carboxylate, Methyl 6-chloropicolinate

M2640-1G M2640-5G
TCIAM2640-1GEA 16 EUR
TCIAM2640-1G TCIAM2640-5G
Methyl-6-chloropicolinate ≥98.0% (by GC)
Methyl-6-chloropicolinate
Formula: C₇H₆ClNO₂
Peso molecolare: 171,58 g/mol
MDL Number: MFCD00155391
CAS Number: 6636-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e7b3ae647d3ac1b2246fea2a76e79e66baa51a2&vid=4e7b3ae647d3ac1b2246fea2a76e79e66baa51a2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR