Stampa…

Methyl 4-(Phenylcarbamoyl)benzoate ≥98.0% (by GC)

Fornitore: TCI

M2637-1G M2637-5G
TCIAM2637-1GEA 24 EUR
TCIAM2637-1G TCIAM2637-5G
Methyl 4-(Phenylcarbamoyl)benzoate ≥98.0% (by GC)
Methyl 4-(Phenylcarbamoyl)benzoate
Formula: C₁₅H₁₃NO₃
MDL Number: MFCD00157832
CAS Number: 3814-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dafa873261860c310c9fb08580d686d00b185256&vid=dafa873261860c310c9fb08580d686d00b185256&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 192.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR