2-Hydroxyethylmesitylene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₆O Punto di ebollizione: 152°C |
MDL Number:
MFCD00020609 CAS Number: 6950-92-1 EINECS: 230-123-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...