Stampa…

4-Methoxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Methoxybenzimidamide hydrochloride

M2612-1G M2612-5G
TCIAM2612-1GEA 68 EUR
TCIAM2612-1G TCIAM2612-5G
4-Methoxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Methoxybenzamidine hydrochloride
Formula: C₈H₁₀N₂O
Peso molecolare: 186,64 g/mol
MDL Number: MFCD00466114
CAS Number: 51721-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ebec8cb125470ff639bc1afe9b9d5439041775&vid=22ebec8cb125470ff639bc1afe9b9d5439041775&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 219.0 to 223.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR