Stampa…

N-Methyl-N-(trifluoromethylthio)aniline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

M2595-200MG M2595-1G
TCIAM2595-200MGEA 114 EUR
TCIAM2595-200MG TCIAM2595-1G
N-Methyl-N-(trifluoromethylthio)aniline ≥98.0% (by GC)
N-Methyl-N-(trifluoromethylthio)aniline
Formula: C₈H₈F₃NS
Peso molecolare: 207,21 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 66476-44-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3c97c4d95036b2513dbdacddb191ecba7494adf&vid=c3c97c4d95036b2513dbdacddb191ecba7494adf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR