Methyl-4-bromo-1H-pyrrole-2-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 4-bromopyrrole-2-carboxylate, Methyl 4-bromo-1H-pyrrole-2-carboxylate
M2583-1G
M2583-5G
TCIAM2583-1GEA
40
EUR
TCIAM2583-1G
TCIAM2583-5G
Methyl-4-bromo-1H-pyrrole-2-carboxylate ≥98.0% (by GC)
Methyl-4-bromo-1H-pyrrole-2-carboxylate
Formula:
C₆H₆BrNO₂ Peso molecolare: 204,00 g/mol |
MDL Number:
MFCD00832855 CAS Number: 934-05-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...