Stampa…

2-(Methylthio)acetamide ≥98.0% (by GC)

Fornitore: TCI
Attenzione

M2579-1G M2579-5G
TCIAM2579-1GEA 16 EUR
TCIAM2579-1G TCIAM2579-5G
2-(Methylthio)acetamide ≥98.0% (by GC)
2-(Methylthio)acetamide
Formula: C₃H₇NOS
MDL Number: MFCD00041321
CAS Number: 22551-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48f374ca41907d70428ffb74b359c3b820f32ddd&vid=48f374ca41907d70428ffb74b359c3b820f32ddd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR