Stampa…

4-Methoxyphenylurea ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-Methoxyphenyl)urea

M2575-1G M2575-5G
TCIAM2575-1GEA 40 EUR
TCIAM2575-1G TCIAM2575-5G
4-Methoxyphenylurea ≥98.0% (by HPLC, total nitrogen)
4-Methoxyphenylurea
Formula: C₈H₁₀N₂O₂
Peso molecolare: 166,18 g/mol
MDL Number: MFCD00014789
CAS Number: 1566-42-3
EINECS: 216-368-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=599a666f64c87494b4472abb7e1d82cde2d3ec02&vid=599a666f64c87494b4472abb7e1d82cde2d3ec02&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR