Stampa…

N-Methyl-2-(4-methoxyphenyl)ethylamine ≥95.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-[2-(Methylamino)ethyl]anisole, 4-Methoxy-N-methylphenethylamine

M2551-20MG M2551-200MG
TCIAM2551-20MGEA 25 EUR
TCIAM2551-20MG TCIAM2551-200MG
N-Methyl-2-(4-methoxyphenyl)ethylamine ≥95.0% (by GC, titration analysis)
N-Methyl-2-(4-methoxyphenyl)ethylamine
Formula: C₁₀H₁₅NO
Peso molecolare: 165.24 g/mol
Punto di ebollizione: 83°C
MDL Number: MFCD00870496
CAS Number: 4091-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edcd0009d340701ff86955d74917067cddc5d6bf&vid=edcd0009d340701ff86955d74917067cddc5d6bf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR