Stampa…

Methyl isocyanoacetate ≥97.0% (by GC)

Fornitore: TCI
Pericolo

M2549-1G M2549-5G
TCIAM2549-1GEA 24 EUR
TCIAM2549-1G TCIAM2549-5G
Methyl isocyanoacetate ≥97.0% (by GC)
Methyl isocyanoacetate
Formula: C₄H₅NO₂
Punto di ebollizione: 75°C
Flash Pt: 84°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00000006
CAS Number: 39687-95-1
EINECS: 254-593-8
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12108dba9dfc53c7ce36c1d13cb6155b11bc523e&vid=12108dba9dfc53c7ce36c1d13cb6155b11bc523e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR