Stampa…

Methyl-2-methyl-4-nitrobenzoate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 2-methyl-4-nitrobenzoate

M2545-5G M2545-25G
TCIAM2545-5GEA 49 EUR
TCIAM2545-5G TCIAM2545-25G
Methyl-2-methyl-4-nitrobenzoate ≥98.0% (by GC)
Methyl-2-methyl-4-nitrobenzoate
Formula: C₉H₉NO₄
Peso molecolare: 195,1700 g/mol
MDL Number: MFCD03618407
CAS Number: 62621-09-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4eb58f32d50aa51f9ffd78819caf4619aab9b5ed&vid=4eb58f32d50aa51f9ffd78819caf4619aab9b5ed&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR