Stampa…

Methyl-5-bromo-2-methylbenzoate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 5-bromo-2-methylbenzoate, 5-Bromo-2-methylbenzoic acid methyll ester, 5-Bromo-o-toluic acid methyll ester, Methyl 5-Bromo-o-toluate, methyl 5-bromo-2-methyl-benzoate

M2542-1G M2542-5G
TCIAM2542-1GEA 17 EUR
TCIAM2542-1G TCIAM2542-5G
Methyl-5-bromo-2-methylbenzoate ≥98.0% (by GC)
Methyl-5-bromo-2-methylbenzoate
Formula: C₉H₉BrO₂
Peso molecolare: 229,073 g/mol
Punto di ebollizione: 265°C
MDL Number: MFCD09839555
CAS Number: 79669-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb44f6d1141a011c024880fbcaf2ec03e32b1e87&vid=bb44f6d1141a011c024880fbcaf2ec03e32b1e87&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR