Stampa…

Methyl-2-amino-5-methylbenzoate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 2-amino-5-methylbenzoate

M2541-5G M2541-25G
TCIAM2541-5GEA 49 EUR
TCIAM2541-5G TCIAM2541-25G
Methyl-2-amino-5-methylbenzoate ≥98.0% (by GC, titration analysis)
Methyl-2-amino-5-methylbenzoate
Formula: C₉H₁₁NO₂
Peso molecolare: 165,1900 g/mol
MDL Number: MFCD00661547
CAS Number: 18595-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fff1546a4122cf53fcfbe2363d4a88b067da4c9&vid=0fff1546a4122cf53fcfbe2363d4a88b067da4c9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR