Methyl 5-bromo-2-furoate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate
Formula:
C₆H₅BrO₃ Peso molecolare: 205.01 g/mol Punto di ebollizione: 80°C |
MDL Number:
MFCD00092313 CAS Number: 2527-99-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...