Stampa…

2-Methoxyestradiol ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol

M2530-25MG M2530-100MG
TCIAM2530-25MGEA 89 EUR
TCIAM2530-25MG TCIAM2530-100MG
2-Methoxyestradiol ≥97.0% (by GC)
2-Methoxyestradiol
Formula: C₁₉H₂₆O₃
Peso molecolare: 302,408 g/mol
MDL Number: MFCD00010489
CAS Number: 362-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f45599d1fdcdbeaa56b04e75a5d62539c1607fa&vid=6f45599d1fdcdbeaa56b04e75a5d62539c1607fa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 186.0 to 190.0 deg-C
Specific rotation [a]20/D +105 to + 110 deg (C=0.5, THF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR