Stampa…

Maprotiline hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-(gamma-Methylaminopropyl)-9,10-dihydro-9,10-ethanoanthracene Hydrochloride

M2527-1G M2527-5G
TCIAM2527-1GEA 64 EUR
TCIAM2527-1G TCIAM2527-5G
Maprotiline hydrochloride ≥98.0% (by HPLC, titration analysis)
Maprotiline hydrochloride
Formula: C₂₀H₂₃N
Peso molecolare: 313,87 g/mol
MDL Number: MFCD00079464
CAS Number: 10347-81-6
EINECS: 233-758-8
Merck Index: 14,05748
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccb56b84a956fd43c26e4d3ec02fa10e4840fb63&vid=ccb56b84a956fd43c26e4d3ec02fa10e4840fb63&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 240.0 to 244.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR