N1-Methyl-4-nitro-1,2-phenylenediamine ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
M2523-1G
M2523-5G
TCIAM2523-1GEA
30
EUR
TCIAM2523-1G
TCIAM2523-5G
N1-Methyl-4-nitro-1,2-phenylenediamine ≥98.0% (by GC)
N1-Methyl-4-nitro-1,2-phenylenediamine
Formula:
C₇H₉N₃O₂ |
MDL Number:
MFCD00156607 CAS Number: 41939-61-1 UN: 2811 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | Red to Dark red to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 177.0 to 181.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...