Methyl caffeate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,4-Dihydroxycinnamic Acid Methyl Ester, Methyl 3,4-Dihydroxycinnamate, Caffeic Acid Methyl Ester
M2519-200MG
M2519-1G
TCIAM2519-200MGEA
76
EUR
TCIAM2519-200MG
TCIAM2519-1G
Methyl caffeate ≥98.0% (by GC, titration analysis)
Methyl caffeate
Formula:
C₁₀H₁₀O₄ Peso molecolare: 194,19 g/mol |
MDL Number:
MFCD00210468 CAS Number: 67667-67-8 |
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...