Methyl-4-methoxypicolinate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 4-methoxypyridine-2-carboxylate
M2508-1G
M2508-5G
TCIAM2508-1GEA
80
EUR
TCIAM2508-1G
TCIAM2508-5G
Methyl-4-methoxypicolinate ≥98.0% (by GC, titration analysis)
Methyl-4-methoxypicolinate
Formula:
C₈H₉NO₃ Peso molecolare: 167,16 g/mol |
MDL Number:
MFCD00956223 CAS Number: 29681-43-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...