Stampa…

5-Nitro-6-methyluracil ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Methyl-5-nitrouracil, 2,4-Dihydroxy-6-methyl-5-nitropyrimidine

M2506-1G M2506-5G
TCIAM2506-1GEA 26 EUR
TCIAM2506-1G TCIAM2506-5G
5-Nitro-6-methyluracil ≥95.0% (by HPLC, titration analysis)
5-Nitro-6-methyluracil
Formula: C5H5N3O4
Peso molecolare: 171,11 g/mol
MDL Number: MFCD00051201
CAS Number: 16632-21-6
EINECS: 240-688-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR