2-Methyl-5-oxo-1-cyclopentenyl propionate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Propionic Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
M2487-5G
M2487-25G
TCIAM2487-5GEA
100
EUR
TCIAM2487-5G
TCIAM2487-25G
2-Methyl-5-oxo-1-cyclopentenyl propionate ≥98.0% (by GC)
2-Methyl-5-oxo-1-cyclopentenyl propionate
Formula:
C₉H₁₂O₃ Peso molecolare: 168,19 g/mol |
MDL Number:
MFCD00798053 CAS Number: 87-55-8 EINECS: 201-751-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...