Stampa…

2,2'-(Methylimino)bis(N,N-di-n-octylacetamide) ≥95.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: MIDOA

M2476-1ML M2476-5ML
TCIAM2476-1MLEA 67 EUR
TCIAM2476-1ML TCIAM2476-5ML
2,2'-(Methylimino)bis(N,N-di-n-octylacetamide) ≥95.0% (by GC, titration analysis)
2,2'-(Methylimino)bis(N,N-di-n-octylacetamide)
Formula: C₃₇H₇₅N₃O₂
Peso molecolare: 594,03 g/mol
MDL Number: MFCD28386110
CAS Number: 1000668-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b7496d61f8aef7c20468f706ab119a139266ea7&vid=1b7496d61f8aef7c20468f706ab119a139266ea7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR