4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers) ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,2-Propyleneglycol sulphite, 1,2-Propanediol Cyclic Sulphite, 1,2-Propylene sulphite
M2471-5G
M2471-25G
TCIAM2471-5GEA
29
EUR
TCIAM2471-5G
TCIAM2471-25G
4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers) ≥98.0% (by GC)
4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers)
Formula:
C₃H₆O₃S Peso molecolare: 122,14 g/mol Punto di ebollizione: 40°C Flash Pt: 81°C |
MDL Number:
MFCD00041209 CAS Number: 1469-73-4 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...