Stampa…

Methyl-5-methylpyrazole-3-carboxylate ≥98.0% (by GC)

Fornitore: TCI

Synonyms: Methyl 5-methyl-1H-pyrazole-3-carboxylate, Methyl 5-methyl[1H]pyrazole-3-carboxylate, 5-Methylpyrazole-3-carboxylic Acid Methyl Ester

M2465-1G M2465-5G
TCIAM2465-1GEA 58 EUR
TCIAM2465-1G TCIAM2465-5G
Methyl-5-methylpyrazole-3-carboxylate ≥98.0% (by GC)
Methyl-5-methylpyrazole-3-carboxylate
Formula: C₆H₈N₂O₂
Peso molecolare: 140.14 g/mol
MDL Number: MFCD03778987
CAS Number: 25016-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f92d75d5565d6bd5ba046853d2f26e406b274b9&vid=9f92d75d5565d6bd5ba046853d2f26e406b274b9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR