Stampa…

Miltefosine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Hexadecyl 2-(Trimethylammonio)ethyl phosphate, Phosphoric acid hexadecyl 2-(trimethylammonio)ethyl ester, Fos-Choline™ 16, HePC, N-Hexadecylphosphocholine, Hexadecyl Phosphorylcholine

M2445-100MG M2445-1G
TCIAM2445-100MGEA 24 EUR
TCIAM2445-100MG TCIAM2445-1G
Miltefosine ≥98.0% (by HPLC, titration analysis)
Miltefosine
Formula: C₂₁H₄₆NO₄P
Peso molecolare: 407,57 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00133396
CAS Number: 58066-85-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,06198
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c1e76f54322368defef9219ba82b941fd9f1b2a&vid=4c1e76f54322368defef9219ba82b941fd9f1b2a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR