Stampa…

1-Methyl-3-octylimidazolium trifluoromethansulphonate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methyl-3-n-octylimidazolium trifluoromethanesulphonate, 1-Methyl-3-n-octylimidazolium triflate, 1-Methyl-3-n-octylimidazolium Trifluoromethanesulphonate

M2440-5G M2440-25G
TCIAM2440-5GEA 79 EUR
TCIAM2440-5G TCIAM2440-25G
1-Methyl-3-octylimidazolium trifluoromethansulphonate ≥98.0% (by HPLC, total nitrogen)
1-Methyl-3-octylimidazolium trifluoromethansulphonate
Formula: C₁₃H₂₃F₃N₂O₃S
Peso molecolare: 344,39 g/mol
MDL Number: MFCD06411717
CAS Number: 403842-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bea51c1168cebcd507f7f08fdf948a4dc46cf1c&vid=8bea51c1168cebcd507f7f08fdf948a4dc46cf1c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR