Stampa…

N-Methyl-L-alanine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: H-N-Me-Ala-OH, N-Me-Ala-OH

M2436-1G M2436-5G
TCIAM2436-1GEA 31 EUR
TCIAM2436-1G TCIAM2436-5G
N-Methyl-L-alanine ≥98.0% (by titrimetric analysis)
N-Methyl-L-alanine
Formula: C₄H₉NO₂
Peso molecolare: 103,12 g/mol
MDL Number: MFCD00037241
CAS Number: 3913-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=856453810e4b1efff3400c4244eb5e5cc8ebaa0d&vid=856453810e4b1efff3400c4244eb5e5cc8ebaa0d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +10.0 to +12.0 deg (C=10, 6mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR