4-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Hydroxy-4-methylbenzaldehyde
M2431-5G
M2431-25G
TCIAM2431-5GEA
94
EUR
TCIAM2431-5G
TCIAM2431-25G
4-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
4-Methylsalicylaldehyde
Formula:
C₈H₈O₂ Peso molecolare: 136,15 g/mol Punto di ebollizione: 223°C |
MDL Number:
MFCD00799550 CAS Number: 698-27-1 EINECS: 211-813-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...