Methyl-5-methylnicotinate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 5-methylnicotinate
M2428-1G
M2428-5G
TCIAM2428-1GEA
27
EUR
TCIAM2428-1G
TCIAM2428-5G
Methyl-5-methylnicotinate ≥98.0% (by GC, titration analysis)
Methyl-5-methylnicotinate
Formula:
C₈H₉NO₂ Peso molecolare: 151,16 g/mol Punto di ebollizione: 90°C |
MDL Number:
MFCD01075739 CAS Number: 29681-45-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...