Stampa…

7-Methoxy-1-naphthylacetonitrile ≥99.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 7-Methoxy-1-naphthylmethylcyanide

M2421-1G M2421-5G
TCIAM2421-1GEA 25 EUR
TCIAM2421-1G TCIAM2421-5G
7-Methoxy-1-naphthylacetonitrile ≥99.0% (by GC)
7-Methoxy-1-naphthylacetonitrile
Formula: C₁₃H₁₁NO
Peso molecolare: 197,24 g/mol
MDL Number: MFCD08704309
CAS Number: 138113-08-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c98eb86b621df3d0cc55ae40aed7a6ba2541a0b&vid=0c98eb86b621df3d0cc55ae40aed7a6ba2541a0b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 82.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR