4-Mercaptophenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₇BO₂S Punto di fusione: 220°C |
MDL Number:
MFCD03093887 CAS Number: 237429-33-3 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...