Stampa…

3-Mercaptophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

M2417-1G M2417-5G
TCIAM2417-1GEA 138 EUR
TCIAM2417-1G TCIAM2417-5G
3-Mercaptophenylboronic acid (contains varying amounts of Anhydride)
3-Mercaptophenylboronic acid
Formula: C₆H₇BO₂S
Punto di fusione: 201°C
MDL Number: MFCD03095131
CAS Number: 352526-01-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7106e82d67bbf83fb442e892d9bdfb51297a5e83&vid=7106e82d67bbf83fb442e892d9bdfb51297a5e83&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR