4-Methyl-2-hexylamine hydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methylhexanamine Hydrochloride, 2-Amino-4-methylhexane Hydrochloride, 1,3-Dimethylamylamine Hydrochloride, 1,3-Dimethylpentylamine Hydrochloride
M2409-1G
M2409-5G
M2409-25G
TCIAM2409-1GEA
18
EUR
TCIAM2409-1G
TCIAM2409-5G
TCIAM2409-25G
4-Methyl-2-hexylamine hydrochloride ≥98.0% (by titrimetric analysis)
4-Methyl-2-hexylamine hydrochloride
Formula:
C₇H₁₇N Peso molecolare: 151,68 g/mol |
MDL Number:
MFCD09038354 CAS Number: 13803-74-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...