Stampa…

Mecarbinate ≥98.0% (by GC)

Fornitore: TCI

M2407-1G M2407-5G
TCIAM2407-1GEA 76 EUR
TCIAM2407-1G TCIAM2407-5G
Mecarbinate ≥98.0% (by GC)
Mecarbinate
Formula: C₁₃H₁₅NO₃
MDL Number: MFCD00451373
CAS Number: 15574-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd3494c81267a9f2e40a7eca20c773af2bc2df95&vid=fd3494c81267a9f2e40a7eca20c773af2bc2df95&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 210.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR