Stampa…

5-Methylisoxazole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

M2405-5G M2405-25G
TCIAM2405-5GEA 65 EUR
TCIAM2405-5G TCIAM2405-25G
5-Methylisoxazole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Methylisoxazole-3-carboxylic acid
Formula: C₅H₅NO₃
Peso molecolare: 127,1 g/mol
Punto di fusione: 174°C
MDL Number: MFCD01318162
CAS Number: 3405-77-4
EINECS: 222-289-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=353fffcb05e67110e3d1e152e5cc7a9356d15709&vid=353fffcb05e67110e3d1e152e5cc7a9356d15709&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR