Stampa…

2-(4-Methoxybenzoyl)thiophene ≥98.0% (by GC)

Fornitore: TCI

M2395-5G M2395-25G
TCIAM2395-5GEA 30 EUR
TCIAM2395-5G TCIAM2395-25G
2-(4-Methoxybenzoyl)thiophene ≥98.0% (by GC)
2-(4-Methoxybenzoyl)thiophene
Formula: C₁₂H₁₀O₂S
MDL Number: MFCD00005440
CAS Number: 4160-63-8
EINECS: 223-997-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c2e02865d075bb45ae23a17cf10d45849e715f5&vid=3c2e02865d075bb45ae23a17cf10d45849e715f5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR