Stampa…

2-(4-Methoxycyclohexyl)ethylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-(2-Aminoethyl)-4-methoxycyclohexane

M2381-1G M2381-5G
TCIAM2381-1GEA 187 EUR
TCIAM2381-1G TCIAM2381-5G
2-(4-Methoxycyclohexyl)ethylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
2-(4-Methoxycyclohexyl)ethylamine (cis- and trans- mixture)
Formula: C₉H₁₉NO
Peso molecolare: 157,26 g/mol
MDL Number: MFCD07772981
CAS Number: 1052223-70-7
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01bd0534229b736de2294be5df84f697bb40893b&vid=01bd0534229b736de2294be5df84f697bb40893b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR