Stampa…

4-Methylpelargonic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

M2380-5ML M2380-25ML
TCIAM2380-5MLEA 40 EUR
TCIAM2380-5ML TCIAM2380-25ML
4-Methylpelargonic acid ≥97.0% (by GC, titration analysis)
4-Methylpelargonic acid
Formula: C₁₀H₂₀O₂
Punto di ebollizione: 145°C
MDL Number: MFCD00051789
CAS Number: 45019-28-1
EINECS: 256-180-8
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e1d6bd5203c48c2382d14439517240698a2c1a1&vid=4e1d6bd5203c48c2382d14439517240698a2c1a1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR