5-Methoxyflavone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
5-Methoxy-2-phenylchromen-4-one, 5-Methoxy-2-phenyl-4H-chromen-4-one, 5-Methoxy-2-phenyl-4-benzopyrone, 5-Methoxy-2-phenyl-4H-1-benzopyran-4-one, 5-Methoxy-2-phenyl-4H-benzo[b]pyran-4-one
M2365-1G
M2365-5G
TCIAM2365-1GEA
80
EUR
TCIAM2365-1G
TCIAM2365-5G
5-Methoxyflavone ≥98.0% (by GC)
5-Methoxyflavone
Formula:
C₁₆H₁₂O₃ Peso molecolare: 252,27 g/mol |
MDL Number:
MFCD00016942 CAS Number: 42079-78-7 EINECS: 255-652-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...