Stampa…

3-Methoxyflavone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3-Methoxy-2-phenyl-4H-chromen-4-one

M2364-1G M2364-5G
TCIAM2364-1GEA 90 EUR
TCIAM2364-1G TCIAM2364-5G
3-Methoxyflavone ≥98.0% (by GC)
3-Methoxyflavone
Formula: C₁₆H₁₂O₃
Peso molecolare: 252,27 g/mol
MDL Number: MFCD00017612
CAS Number: 7245-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f804413a51d0a00e2fdb7efaae1b6772e0acb9c&vid=0f804413a51d0a00e2fdb7efaae1b6772e0acb9c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR