Myricitrin ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
3,3',4',5,5',7-Hexahydroxyflavone 3-O-Rhamnoside
M2361-10MG
M2361-50MG
TCIAM2361-10MGEA
57
EUR
TCIAM2361-10MG
TCIAM2361-50MG
Myricitrin ≥98.0% (by HPLC)
Myricitrin
Formula:
C₂₁H₂₀O₁₂ Peso molecolare: 464,38 g/mol Punto di fusione: 197°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00016930 CAS Number: 17912-87-7 EINECS: 241-856-7 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -149 to -154 deg (C=0.5、MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...