3-[[2-(Methacryloyloxy)ethyl]dimethylammonio]propionate ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
3-((2-(Methacryloyloxy)ethyl)dimethylammonio)propanoate
M2359-1G
M2359-5G
TCIAM2359-1GEA
80
EUR
TCIAM2359-1G
TCIAM2359-5G
3-[[2-(Methacryloyloxy)ethyl]dimethylammonio]propionate ≥98.0% (by HPLC)
3-[[2-(Methacryloyloxy)ethyl]dimethylammonio]propionate
Formula:
C₁₁H₁₉NO₄ Peso molecolare: 229,28 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD28386109 CAS Number: 24249-95-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...