Methyl-1-aminocyclopentanecarboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
1-Aminocyclopentanecarboxylic Acid Methyl Ester Hydrochloride, Methyl 1-aminocyclopentanecarboxylate hydrochloride, Methyl 1-Aminocyclopentanecarboxylate HCl, Cycloleucine Methyl Ester Hydrochloride
M2351-1G
M2351-5G
TCIAM2351-1GEA
20
EUR
TCIAM2351-1G
TCIAM2351-5G
Methyl-1-aminocyclopentanecarboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Methyl-1-aminocyclopentanecarboxylate hydrochloride
Formula:
C₇H₁₃NO₂ Peso molecolare: 179,64 g/mol |
MDL Number:
MFCD03840364 CAS Number: 60421-23-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...