Stampa…

3-Formyl-6-methoxypyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde

M2347-1G M2347-5G
TCIAM2347-1GEA 38 EUR
TCIAM2347-1G TCIAM2347-5G
3-Formyl-6-methoxypyridine ≥98.0% (by GC)
3-Formyl-6-methoxypyridine
Formula: C₇H₇NO₂
Peso molecolare: 137,14 g/mol
MDL Number: MFCD02683446
CAS Number: 65873-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44163cc36558aea85f2135e5a480365b24d385af&vid=44163cc36558aea85f2135e5a480365b24d385af&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR