Stampa…

6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 6-Methoxy-2-(4-methoxyphenyl)-1-benzothiophene

M2345-5G M2345-25G
TCIAM2345-5GEA 54 EUR
TCIAM2345-5G TCIAM2345-25G
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene ≥98.0% (by GC)
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene
Formula: C₁₆H₁₄O₂S
Peso molecolare: 270,35 g/mol
MDL Number: MFCD02083138
CAS Number: 63675-74-1
EINECS: 264-408-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25982940136f3ccade809b8202de7d47dd43ed6f&vid=25982940136f3ccade809b8202de7d47dd43ed6f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR