6-Methyl-3(2H)-pyridazinone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Methylpyridazin-3(2H)-one
M2344-1G
M2344-5G
TCIAM2344-1GEA
16
EUR
TCIAM2344-1G
TCIAM2344-5G
6-Methyl-3(2H)-pyridazinone ≥98.0% (by GC)
6-Methyl-3(2H)-pyridazinone
Formula:
C₅H₆N₂O Peso molecolare: 110,12 g/mol |
MDL Number:
MFCD00039720 CAS Number: 13327-27-0 EINECS: 236-367-0 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...