Stampa…

Methyl-6-bromopicolinate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 6-bromopicolinate, Methyl 6-bromopyridine-2-carboxylate, 6-Bromopyridine-2-carboxylic acid methyl ester, 6-Bromopicolinic acid methyl ester

M2342-1G M2342-5G
TCIAM2342-1GEA 16 EUR
TCIAM2342-1G TCIAM2342-5G
Methyl-6-bromopicolinate ≥98.0% (by GC)
Methyl-6-bromopicolinate
Formula: C₇H₆BrNO₂
Peso molecolare: 216,03 g/mol
MDL Number: MFCD06203934
CAS Number: 26218-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d04fa2126fa6a76afbc31fe4b5fd75a273dc1af&vid=7d04fa2126fa6a76afbc31fe4b5fd75a273dc1af&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR